D-erythro-Sphingosine hydrochloride, 97%
D-erythro-Sphingosine hydrochloride, 97%
D-erythro-Sphingosine hydrochloride, 97%
D-erythro-Sphingosine hydrochloride, 97%
Thermo Scientific Chemicals

D-erythro-Sphingosine hydrochloride, 97%

CAS: 2673-72-5 | C18H38ClNO2 | 335.96 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 mg
500 mg
Catalog number H52427.ME
also known as H52427-ME
Price (USD)
1,054.65
Special offer
Online exclusive
Ends: 30-Jun-2026
1,241.00
Save 186.35 (15%)
Each
Add to cart
Quantity:
500 mg
Request bulk or custom format
Price (USD)
1,054.65
Special offer
Online exclusive
Ends: 30-Jun-2026
1,241.00
Save 186.35 (15%)
Each
Add to cart
Specifications
CAS2673-72-5
Chemical Name or MaterialD-erythro-Sphingosine hydrochloride
Formula Weight335.96
IUPAC Name(E,2S,3R)-2-aminooctadec-4-ene-1,3-diol;hydrochloride
InChI KeyYDIHJJLAPMAISR-UHFFFAOYNA-N
View more
Inhibitor of protein kinase C and calmodulin-dependent enzymes, but may stimulate mast cells by activation of protein kinase C

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only