N-Boc-3-hydroxy-L-valine, 97%
N-Boc-3-hydroxy-L-valine, 97%
N-Boc-3-hydroxy-L-valine, 97%
N-Boc-3-hydroxy-L-valine, 97%
Thermo Scientific Chemicals

N-Boc-3-hydroxy-L-valine, 97%

CAS: 102507-13-1 | C10H19NO5 | 233.26 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 mg
1 g
Catalog number H52429.MD
also known as H52429-MD
Price (USD)
297.65
Online Exclusive
331.00
Save 33.35 (10%)
Each
Add to cart
Quantity:
250 mg
Request bulk or custom format
Price (USD)
297.65
Online Exclusive
331.00
Save 33.35 (10%)
Each
Add to cart
Specifications
CAS102507-13-1
Chemical Name or MaterialN-Boc-3-hydroxy-L-valine
Formula Weight233.26
IUPAC Name(2S)-3-hydroxy-3-methyl-2-[(2-methylpropan-2-yl)oxycarbonylamino]butanoic acid
InChI KeySZVRVSZFEDIMFM-ZCFIWIBFSA-N
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only