2-(Bromomethyl)benzeneboronic acid, 97%
2-(Bromomethyl)benzeneboronic acid, 97%
2-(Bromomethyl)benzeneboronic acid, 97%
Thermo Scientific Chemicals

2-(Bromomethyl)benzeneboronic acid, 97%

CAS: 91983-14-1 | C7H8BBrO2 | 214.85 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number H52467.06
also known as H52467-06
Price (USD)
308.65
Special offer
Online exclusive
Ends: 30-Jun-2026
363.00
Save 54.35 (15%)
Each
Quantity:
5 g
Request bulk or custom format
Price (USD)
308.65
Special offer
Online exclusive
Ends: 30-Jun-2026
363.00
Save 54.35 (15%)
Each
Chemical Identifiers
CAS91983-14-1
IUPAC Name[2-(bromomethyl)phenyl]boronic acid
Molecular FormulaC7H8BBrO2
InChI KeyMYVJCOQGXCONPE-UHFFFAOYSA-N
SMILESOB(O)C1=CC=CC=C1CBr
View more
SpecificationsSpecification SheetSpecification Sheet
FormPowder
Appearance (Color)White
Assay (HPLC)≥96.0%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only