Specifications
CAS913835-67-3
Chemical Name or Material1-Boc-6-cyanoindole-2-boronic acid
Formula Weight286.09
Health Hazard 1H302-H315-H319-H335
Health Hazard 2GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
View more