1-Boc-6-cyanoindole-2-boronic acid, 96%
1-Boc-6-cyanoindole-2-boronic acid, 96%
1-Boc-6-cyanoindole-2-boronic acid, 96%
1-Boc-6-cyanoindole-2-boronic acid, 96%
Thermo Scientific Chemicals

1-Boc-6-cyanoindole-2-boronic acid, 96%

CAS: 913835-67-3 | C14H15BN2O4 | 286.09 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 mg
1 g
Catalog number H52520.MD
also known as H52520-MD
Price (USD)
58.70
Each
Add to cart
Quantity:
250 mg
Request bulk or custom format
Price (USD)
58.70
Each
Add to cart
Specifications
CAS913835-67-3
Chemical Name or Material1-Boc-6-cyanoindole-2-boronic acid
Formula Weight286.09
Health Hazard 1H302-H315-H319-H335
Health Hazard 2GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only