3-(2-Methoxycarbonylethyl)benzeneboronic acid, 97%
3-(2-Methoxycarbonylethyl)benzeneboronic acid, 97%
3-(2-Methoxycarbonylethyl)benzeneboronic acid, 97%
Thermo Scientific Chemicals

3-(2-Methoxycarbonylethyl)benzeneboronic acid, 97%

CAS: 833472-82-5 | C10H13BO4 | 208.02 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 mg
1 g
Catalog number H52542.03
also known as H52542-03
Price (USD)
261.00
Each
Quantity:
1 g
Request bulk or custom format
Price (USD)
261.00
Each
Specifications
CAS833472-82-5
Chemical Name or Material3-(2-Methoxycarbonylethyl)benzeneboronic Acid
Formula Weight208.02
Health Hazard 1H315-H319-H335
Health Hazard 2GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
View more
3-(2-Methoxycarbonylethyl)benzeneboronic acid is involved in the preparation of antimalarial and anticancer ethylenediamine inhibitors to protein farnesyltransferase.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3-(2-Methoxycarbonylethyl)benzeneboronic acid is involved in the preparation of antimalarial and anticancer ethylenediamine inhibitors to protein farnesyltransferase.

Notes
Incompatible with strong oxidizing agents and strong acids.
RUO – Research Use Only
Hast, M. A.; Steven, F.; Christopher, G. C.; Erin, E. P.; Michelle, A. B.; Kasey, R.; Michael, H. G.; Wesley, C. V. V.; Said, M. S.; Andrew, D. H.; Beese, L. S. Structural Basis for Binding and Selectivity of Antimalarial and Anticancer Ethylenediamine Inhibitors to Protein Farnesyltransferase. Chem. Biol. 2009, 16 (2), 181-192.