O-Methyl-L-serine hydrochloride, 95%
O-Methyl-L-serine hydrochloride, 95%
O-Methyl-L-serine hydrochloride, 95%
O-Methyl-L-serine hydrochloride, 95%
Thermo Scientific Chemicals

O-Methyl-L-serine hydrochloride, 95%

CAS: 336100-47-1 | C4H10ClNO3 | 155.58 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 mg
1 g
Catalog number H52562.03
also known as H52562-03
Price (USD)
418.65
Online Exclusive
465.00
Save 46.35 (10%)
Each
Add to cart
Quantity:
1 g
Request bulk or custom format
Price (USD)
418.65
Online Exclusive
465.00
Save 46.35 (10%)
Each
Add to cart
Chemical Identifiers
CAS336100-47-1
IUPAC Name2-amino-3-methoxypropanoic acid hydrochloride
Molecular FormulaC4H10ClNO3
InChI KeyNLDVBPFPOCWZCE-UHFFFAOYNA-N
SMILESCl.COCC(N)C(O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (unspecified)≥94.0%
FormSolid
Appearance (Color)White to off-white
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only