Specifications
CAS73397-27-0
Chemical Name or Material(2S,3S)-3-(Boc-amino)-2-hydroxy-5-methylhexanoic acid
Formula Weight261.32
Health Hazard 1H315-H319-H335
Health Hazard 2GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
View more