5-Amino-2-(hydroxymethyl)benzeneboronic acid hemiester hydrochloride, 95%
5-Amino-2-(hydroxymethyl)benzeneboronic acid hemiester hydrochloride, 95%
5-Amino-2-(hydroxymethyl)benzeneboronic acid hemiester hydrochloride, 95%
Thermo Scientific Chemicals

5-Amino-2-(hydroxymethyl)benzeneboronic acid hemiester hydrochloride, 95%

CAS: 117098-93-8 | C7H9BClNO2 | 185.41 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 mg
1 g
Catalog number H52629.MD
also known as H52629-MD
Price (USD)
188.65
Online Exclusive
209.00
Save 20.35 (10%)
Each
Quantity:
250 mg
Request bulk or custom format
Price (USD)
188.65
Online Exclusive
209.00
Save 20.35 (10%)
Each
Chemical Identifiers
CAS117098-93-8
SpecificationsSpecification SheetSpecification Sheet
FormCrystals or powder or crystalline powder
Identification (FTIR)Conforms
Appearance (Color)Yellow to gray
Assay (HPLC)≥94.0%
5-Amino-2-(hydroxymethyl)benzeneboronic acid hemiester hydrochloride is used as an organic chemical synthesis intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
5-Amino-2-(hydroxymethyl)benzeneboronic acid hemiester hydrochloride is used as an organic chemical synthesis intermediate.

Notes
Stable under recommended storage conditions. Store away from oxidizing agents.
RUO – Research Use Only