(+/-)-1-Boc-3-(hydroxymethyl)piperidine, 97%
(+/-)-1-Boc-3-(hydroxymethyl)piperidine, 97%
(+/-)-1-Boc-3-(hydroxymethyl)piperidine, 97%
(+/-)-1-Boc-3-(hydroxymethyl)piperidine, 97%
Thermo Scientific Chemicals

(+/-)-1-Boc-3-(hydroxymethyl)piperidine, 97%

CAS: 116574-71-1 | C11H21NO3 | 215.293 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Catalog number H52784.06
also known as H52784-06
Price (USD)
87.65
Online Exclusive
97.50
Save 9.85 (10%)
Each
Add to cart
Quantity:
5 g
Request bulk or custom format
Price (USD)
87.65
Online Exclusive
97.50
Save 9.85 (10%)
Each
Add to cart
Specifications
CAS116574-71-1
Chemical Name or Material(+/-)-1-Boc-3-(hydroxymethyl)piperidine
Formula Weight215.29
Health Hazard 1H315-H319-H335
Health Hazard 2GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
View more
(±)-1-Boc-3-(hydroxymethyl)piperidine is used as a reactant for synthesis of Pim-1 inhibitors, Vasopressin1b receptor antagonists, CXCR4 antagonists as anti-HIV agents, Amide CCR5 antagonist, PSSRI-based inhibitors of S. aureus multidrug efflux pumps and Human GnRH receptor antagonists.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
(+/-)-1-Boc-3-(hydroxymethyl)piperidine is used as a reactant for synthesis of Pim-1 inhibitors, Vasopressin1b receptor antagonists, CXCR4 antagonists as anti-HIV agents, Amide CCR5 antagonist, PSSRI-based inhibitors of S. aureus multidrug efflux pumps and Human GnRH receptor antagonists.

Solubility
Soluble in methanol and dimethylformamide(DMF).

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
RUO – Research Use Only
Peter Beak; Won Koo Lee.alpha.-Lithioamine synthetic equivalents: syntheses of diastereoisomers from Boc derivatives of cyclic amines. J. Org. Chem. 1993, 58, (5),1109-1117