(S)-3-Boc-thiazolidine-2-carboxylic acid, 97%
(S)-3-Boc-thiazolidine-2-carboxylic acid, 97%
(S)-3-Boc-thiazolidine-2-carboxylic acid, 97%
(S)-3-Boc-thiazolidine-2-carboxylic acid, 97%
Thermo Scientific Chemicals

(S)-3-Boc-thiazolidine-2-carboxylic acid, 97%

CAS: 891192-95-3 | C9H15NO4S | 233.282 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number H52793.06
also known as H52793-06
Price (USD)
445.65
Special offer
Online exclusive
Ends: 30-Jun-2026
524.00
Save 78.35 (15%)
Each
Add to cart
Quantity:
5 g
Request bulk or custom format
Price (USD)
445.65
Special offer
Online exclusive
Ends: 30-Jun-2026
524.00
Save 78.35 (15%)
Each
Add to cart
Specifications
CAS891192-95-3
Chemical Name or Material(S)-3-Boc-thiazolidine-2-carboxylic acid
Formula Weight233.28
Health Hazard 1H315-H319-H335
Health Hazard 2GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
View more
(S)-3-Boc-thiazolidine-2-carboxylic acid is used as an organic chemical synthesis intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
(S)-3-Boc-thiazolidine-2-carboxylic acid is used as an organic chemical synthesis intermediate.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
RUO – Research Use Only

General References:

  1. Takayoshi Suzuki; Azusa Matsuura; Akiyasu Kouketsu; Shinya Hisakawa; Hidehiko Nakagawa; Naoki Miyata. Design and synthesis of non-hydroxamate histone deacetylase inhibitors: identification of a selective histone acetylating agent. Bioorganic & Medicinal Chemistry. 2005, 13, (13),4332-4342