1-Boc-2-phenylpiperazine, 97%
1-Boc-2-phenylpiperazine, 97%
1-Boc-2-phenylpiperazine, 97%
1-Boc-2-phenylpiperazine, 97%
Thermo Scientific Chemicals

1-Boc-2-phenylpiperazine, 97%

CAS: 886766-60-5 | C15H22N2O2 | 262.353 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 mg
1 g
Catalog number H52797.MD
also known as H52797-MD
Price (USD)
111.65
Online Exclusive
124.00
Save 12.35 (10%)
Each
Add to cart
Quantity:
250 mg
Request bulk or custom format
Price (USD)
111.65
Online Exclusive
124.00
Save 12.35 (10%)
Each
Add to cart
Specifications
CAS886766-60-5
Chemical Name or Material1-Boc-2-phenylpiperazine
Formula Weight262.35
Health Hazard 1H315-H319-H335
Health Hazard 2GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
View more
1-Boc-2-phenylpiperazine is used as a pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-Boc-2-phenylpiperazine is used as a pharmaceutical intermediate.

Solubility
Slightly soluble in water.

Notes
Store in a cool, dry place, in a well sealed container. Store away from oxidizing agents.
RUO – Research Use Only