N(alpha)-Boc-D-2,3-diaminopropionic acid, 97%
N(alpha)-Boc-D-2,3-diaminopropionic acid, 97%
N(alpha)-Boc-D-2,3-diaminopropionic acid, 97%
Thermo Scientific Chemicals

N(alpha)-Boc-D-2,3-diaminopropionic acid, 97%

CAS: 76387-70-7 | C8H16N2O4 | 204.226 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 mg
1 g
5 g
Catalog number H52821.03
also known as H52821-03
Price (USD)
146.65
Online Exclusive
163.00
Save 16.35 (10%)
Each
Quantity:
1 g
Request bulk or custom format
Price (USD)
146.65
Online Exclusive
163.00
Save 16.35 (10%)
Each
Chemical Identifiers
CAS76387-70-7
IUPAC Name3-amino-2-{[(tert-butoxy)carbonyl]amino}propanoic acid
Molecular FormulaC8H16N2O4
InChI KeyKRJLRVZLNABMAT-UHFFFAOYNA-N
SMILESCC(C)(C)OC(=O)NC(CN)C(O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (unspecified)>96.0%
Loss on ignition<0.10%
N(alpha)-Boc-D-2,3-diaminopropionic acid is used as pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
N(alpha)-Boc-D-2,3-diaminopropionic acid is used as pharmaceutical intermediate.

Notes
Stable under recommended storage conditions. Store away from oxidizing agents.
RUO – Research Use Only
Erkin Kuru; Srinivas Tekkam; Edward Hall; Yves V.Brun; Michael S.Van Nieuwenhze. Synthesis of fluorescent D-amino acids and their use for probing peptidoglycan synthesis and bacterial growth in situ. Nature Protocols. 2015, (10),33-52