3-(Dimethylsulfamoyl)benzeneboronic acid, 98%
3-(Dimethylsulfamoyl)benzeneboronic acid, 98%
3-(Dimethylsulfamoyl)benzeneboronic acid, 98%
3-(Dimethylsulfamoyl)benzeneboronic acid, 98%
Thermo Scientific Chemicals

3-(Dimethylsulfamoyl)benzeneboronic acid, 98%

CAS: 871329-59-8 | C8H12BNO4S | 229.057 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 mg
1 g
Catalog number H52838.MD
also known as H52838-MD
Price (USD)
95.65
Special offer
Online exclusive
Ends: 30-Jun-2026
112.00
Save 16.35 (15%)
Each
Add to cart
Quantity:
250 mg
Request bulk or custom format
Price (USD)
95.65
Special offer
Online exclusive
Ends: 30-Jun-2026
112.00
Save 16.35 (15%)
Each
Add to cart
Chemical Identifiers
CAS871329-59-8
IUPAC Name[3-(dimethylsulfamoyl)phenyl]boronic acid
Molecular FormulaC8H12BNO4S
InChI KeySGERXKCDWJPIOS-UHFFFAOYSA-N
SMILESCN(C)S(=O)(=O)C1=CC=CC(=C1)B(O)O
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (HPLC)>97.5%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only