4-n-Propylbenzeneboronic acid, 98%, Thermo Scientific Chemicals
4-n-Propylbenzeneboronic acid, 98%, Thermo Scientific Chemicals
4-n-Propylbenzeneboronic acid, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

4-n-Propylbenzeneboronic acid, 98%, Thermo Scientific Chemicals

Have Questions?
Catalog NumberQuantity
H52884.06
also known as H52884-06
5 g
Catalog number H52884.06
also known as H52884-06
Price (USD)
114.65
Special offer
Online exclusive
Ends: 28-Dec-2026
135.00
Save 20.35
Each
Quantity:
5 g
Request bulk or custom format
Price (USD)
114.65
Special offer
Online exclusive
Ends: 28-Dec-2026
135.00
Save 20.35
Each
Chemical Identifiers
CAS134150-01-9
IUPAC Name(4-propylphenyl)boronic acid
Molecular FormulaC9H13BO2
InChI KeyWLCGYIWOKVWFLB-UHFFFAOYSA-N
SMILESCCCC1=CC=C(C=C1)B(O)O
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (HPLC)>97.5%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only