3-Chloro-4-(cyclopropylcarbamoyl)benzeneboronic acid, 97%, Thermo Scientific Chemicals
3-Chloro-4-(cyclopropylcarbamoyl)benzeneboronic acid, 97%, Thermo Scientific Chemicals
3-Chloro-4-(cyclopropylcarbamoyl)benzeneboronic acid, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

3-Chloro-4-(cyclopropylcarbamoyl)benzeneboronic acid, 97%, Thermo Scientific Chemicals

Have Questions?
Catalog NumberQuantity
H52910.MD
also known as H52910-MD
250 mg
Catalog number H52910.MD
also known as H52910-MD
Price (USD)
93.65
Online Exclusive
104.00
Save 10.35 (10%)
Each
Quantity:
250 mg
Request bulk or custom format
Price (USD)
93.65
Online Exclusive
104.00
Save 10.35 (10%)
Each
Chemical Identifiers
CAS850589-44-5
IUPAC Name[3-chloro-4-(cyclopropylcarbamoyl)phenyl]boronic acid
Molecular FormulaC10H11BClNO3
InChI KeyTUPWHDSMIIRKLY-UHFFFAOYSA-N
SMILESOB(O)C1=CC(Cl)=C(C=C1)C(=O)NC1CC1
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (HPLC)>96.0%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only