4-Acetamidobenzeneboronic acid, 96%
4-Acetamidobenzeneboronic acid, 96%
4-Acetamidobenzeneboronic acid, 96%
4-Acetamidobenzeneboronic acid, 96%
Thermo Scientific Chemicals

4-Acetamidobenzeneboronic acid, 96%

CAS: 101251-09-6 | C8H10BNO3 | 178.98 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number H52987.06
also known as H52987-06
Price (USD)
320.00
Each
Add to cart
Quantity:
5 g
Request bulk or custom format
Price (USD)
320.00
Each
Add to cart
Specifications
CAS101251-09-6
Chemical Name or Material4-Acetamidobenzeneboronic acid
Formula Weight178.98
Health Hazard 1H315-H319-H335
Health Hazard 2GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only