4-Methylnaphthalene-1-boronic acid, 96%
4-Methylnaphthalene-1-boronic acid, 96%
4-Methylnaphthalene-1-boronic acid, 96%
Thermo Scientific Chemicals

4-Methylnaphthalene-1-boronic acid, 96%

CAS: 103986-53-4 | C11H11BO2 | 186.02 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number H52998.03
also known as H52998-03
Price (USD)
38.30
Each
Quantity:
1 g
Request bulk or custom format
Price (USD)
38.30
Each
Chemical Identifiers
CAS103986-53-4
IUPAC Name(4-methylnaphthalen-1-yl)boronic acid
Molecular FormulaC11H11BO2
InChI KeyJHVQEUGNYSVSDH-UHFFFAOYSA-N
SMILESCC1=CC=C(B(O)O)C2=CC=CC=C12
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (HPLC)>95.0%
Reactant for Suzuki-Miyaura coupling.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Reactant for Suzuki-Miyaura coupling.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
RUO – Research Use Only
Green, I. R.; et al. 2-Arylnaphthoquinone analogues: potential anti-TB and pro-apoptotic agents. Journal name. 2011, 192-212.