1-Methylindole-2-boronic acid, 95%
1-Methylindole-2-boronic acid, 95%
1-Methylindole-2-boronic acid, 95%
Thermo Scientific Chemicals

1-Methylindole-2-boronic acid, 95%

CAS: 191162-40-0 | C9H10BNO2 | 174.99 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 mg
1 g
Catalog number H53028.03
also known as H53028-03
Price (USD)
257.00
Each
Quantity:
1 g
Request bulk or custom format
Price (USD)
257.00
Each
Chemical Identifiers
CAS191162-40-0
SpecificationsSpecification SheetSpecification Sheet
Assay (HPLC)>94.0%
Reactant for Suzuki-Miyaura coupling. It ia an active Pharmaceutical ingredient hence used as pharmaceutical intermediates. Also used as rawv material in organic synthesis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Reactant for Suzuki-Miyaura coupling. It ia an active Pharmaceutical ingredient hence used as pharmaceutical intermediates. Also used as rawv material in organic synthesis.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents. Store at 4°C.
RUO – Research Use Only
Green, I. R.; et al. 2-Arylnaphthoquinone analogues: potential anti-TB and pro-apoptotic agents. Journal name. 2011, 192-212.