4-n-Heptyloxybenzeneboronic acid, 97%, Thermo Scientific Chemicals
4-n-Heptyloxybenzeneboronic acid, 97%, Thermo Scientific Chemicals
4-n-Heptyloxybenzeneboronic acid, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

4-n-Heptyloxybenzeneboronic acid, 97%, Thermo Scientific Chemicals

Have Questions?
Catalog NumberQuantity
H53066.06
also known as H53066-06
5 g
Catalog number H53066.06
also known as H53066-06
Price (USD)
103.65
Special offer
Online exclusive
Ends: 30-Jun-2026
122.00
Save 18.35 (15%)
Each
Quantity:
5 g
Request bulk or custom format
Price (USD)
103.65
Special offer
Online exclusive
Ends: 30-Jun-2026
122.00
Save 18.35 (15%)
Each
Chemical Identifiers
CAS136370-19-9
IUPAC Name[4-(heptyloxy)phenyl]boronic acid
Molecular FormulaC13H21BO3
InChI KeyUHRONCCLURKEKO-UHFFFAOYSA-N
SMILESCCCCCCCOC1=CC=C(C=C1)B(O)O
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (HPLC)>96.0%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only