3-(Methylcarbamoyl)benzeneboronic acid, 98%
3-(Methylcarbamoyl)benzeneboronic acid, 98%
3-(Methylcarbamoyl)benzeneboronic acid, 98%
Thermo Scientific Chemicals

3-(Methylcarbamoyl)benzeneboronic acid, 98%

CAS: 832695-88-2 | C8H10BNO3 | 178.98 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 mg
1 g
Catalog number H53141.MD
also known as H53141-MD
Price (USD)
73.65
Online Exclusive
81.80
Save 8.15 (10%)
Each
Quantity:
250 mg
Request bulk or custom format
Price (USD)
73.65
Online Exclusive
81.80
Save 8.15 (10%)
Each
Chemical Identifiers
CAS832695-88-2
IUPAC Name[3-(methylcarbamoyl)phenyl]boronic acid
Molecular FormulaC8H10BNO3
InChI KeyFYFFPNFUVMBPRZ-UHFFFAOYSA-N
SMILESCNC(=O)C1=CC=CC(=C1)B(O)O
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (HPLC)>97.5%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only