4-(Boc-aminomethyl)benzeneboronic acid, 95%
4-(Boc-aminomethyl)benzeneboronic acid, 95%
4-(Boc-aminomethyl)benzeneboronic acid, 95%
Thermo Scientific Chemicals

4-(Boc-aminomethyl)benzeneboronic acid, 95%

CAS: 489446-42-6 | C12H18BNO4 | 251.09 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 mg
1 g
Catalog number H53225.MD
also known as H53225-MD
Price (USD)
67.65
Special offer
Online exclusive
Ends: 30-Jun-2026
79.20
Save 11.55 (15%)
Each
Quantity:
250 mg
Request bulk or custom format
Price (USD)
67.65
Special offer
Online exclusive
Ends: 30-Jun-2026
79.20
Save 11.55 (15%)
Each
Chemical Identifiers
CAS489446-42-6
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White
Assay (HPLC)>94.0%
FormSolid
4-(Boc-aminomethyl)benzeneboronic acid is a boc protected phenyl boronic acid used for proteomics research.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-(Boc-aminomethyl)benzeneboronic acid is a boc protected phenyl boronic acid used for proteomics research.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
RUO – Research Use Only

General References:

  1. Peter D.Williams; Donnette D.Staas; Shankar Venkatraman; H.Marie Loughran; Rowena D.Ruzek; Theresa M.Booth; Terry A.Lyle; John S.Wai; Joseph P.Vacca; Bradley P.Feuston; Linda T.Ecto; Jessica A.Flynn; Daniel J.DiStefano; Daria J.Hazuda; Carolyn M.Bahnck; Amy L.Himmelberger; Geetha Dornadula; Renee C.Hrin; Kara A.Stillmock; Marc V.Witmer; Michael D.Miller; Jay A.Grobler. Potent and selective HIV-1 ribonuclease H inhibitors based on a 1-hydroxy-1,8-naphthyridin-2(1H)-one scaffold. Bioorganic & Medicinal Chemistry Letters. 2010, 20, (22),6754-6757