3-Carboxy-2-methoxybenzeneboronic acid, 98%, Thermo Scientific Chemicals
3-Carboxy-2-methoxybenzeneboronic acid, 98%, Thermo Scientific Chemicals
3-Carboxy-2-methoxybenzeneboronic acid, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

3-Carboxy-2-methoxybenzeneboronic acid, 98%, Thermo Scientific Chemicals

Have Questions?
Catalog NumberQuantity
H53323.03
also known as H53323-03
1 g
Catalog number H53323.03
also known as H53323-03
Price (USD)
197.00
Each
Quantity:
1 g
Request bulk or custom format
Price (USD)
197.00
Each
Specifications
CAS913836-10-9
Chemical Name or Material3-Carboxy-2-methoxybenzeneboronic acid
Formula Weight195.97
Health Hazard 1H315-H319-H335
Health Hazard 2GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only