2-Methyl-1-pentene, 97%
2-Methyl-1-pentene, 97%
2-Methyl-1-pentene, 97%
2-Methyl-1-pentene, 97%
Thermo Scientific Chemicals

2-Methyl-1-pentene, 97%

CAS: 763-29-1 | C6H12 | 84.162 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 mL
25 mL
100 mL
Catalog number H53373.AA
also known as H53373-AA
Price (USD)
49.40
Each
Add to cart
Quantity:
5 mL
Request bulk or custom format
Price (USD)
49.40
Each
Add to cart
Chemical Identifiers
CAS763-29-1
IUPAC Name2-methylpent-1-ene
Molecular FormulaC6H12
InChI KeyWWUVJRULCWHUSA-UHFFFAOYSA-N
SMILESCCCC(C)=C
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (HPLC)96.0% min
2-Methyl-1-pentene is prepared from 2-Methyl-1-pentene. 2-methyl-1-pentene is used in copolymerization of ethylene catalyzed by nonbridged half-titanocenes. The isomerization of 2-methyl-1-pentene leads to anionic activation of c-h bonds in olefins.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Methyl-1-pentene is prepared from 2-Methyl-1-pentene. 2-methyl-1-pentene is used in copolymerization of ethylene catalyzed by nonbridged half-titanocenes. The isomerization of 2-methyl-1-pentene leads to anionic activation of c-h bonds in olefins.

Solubility
Insoluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
RUO – Research Use Only

General References:

  1. Kotohiro Nomura.; Koji Itagaki,; Michiya Fujiki. Efficient Incorporation of 2-Methyl-1-pentene in Copolymerization of Ethylene with 2-Methyl-1-pentene Catalyzed by Nonbridged Half-Titanocenes. Macromolecules. 2005, 38 (6), 2053-2055.
  2. A. Schriesheim,; C. A. Rowe. Anionic Activation of C-H Bonds in Olefins. III. Solvent Effect on the Isomerization of 2-Methyl-1-pentene. J. Am. Chem. Soc., 1962, 84 (16), 3160-3164.