3-(Trimethylsilyl)propiolic acid, 97%
3-(Trimethylsilyl)propiolic acid, 97%
3-(Trimethylsilyl)propiolic acid, 97%
Thermo Scientific Chemicals

3-(Trimethylsilyl)propiolic acid, 97%

CAS: 5683-31-8 | C6H10O2Si | 142.23 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
25 g
Catalog number H53380.14
also known as H53380-14
Price (USD)
833.65
Online Exclusive
926.00
Save 92.35 (10%)
Each
Quantity:
25 g
Request bulk or custom format
Price (USD)
833.65
Online Exclusive
926.00
Save 92.35 (10%)
Each
Chemical Identifiers
CAS5683-31-8
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to pale yellow
Infrared spectrumConforms
FormSolid or liquid
Purity≥96.0%
3-(Trimethylsilyl)propiolic acid may be used in the regioselective preparation of 1,5-trisubstituted 1H-1,2,3-triazoles. It is also used as pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3-(Trimethylsilyl)propiolic acid may be used in the regioselective preparation of 1,5-trisubstituted 1H-1,2,3-triazoles. It is also used as pharmaceutical intermediate.

Solubility
Soluble in methanol.

Notes
Stable under recommended storage conditions. Store away from oxidizing agents.
RUO – Research Use Only

General References:

  1. A.S.Medvedeva; M.V.Andreev; L.P.Safronova. One-Pot Synthesis of 3-(Trimethylsilyl)propynamides. Russian Journal of Organic Chemistry. 2010, 46, (10),1466-1470
  2. Silje Melnes; Annette Bayer; Odd R.Gautun. Aryl N-methyliminodiacetic acid (MIDA) boronates from cyclotrimerization of ethynyl MIDA boronate with diynes. Tetrahedron. 2013, 69, (37),7910-7915