1-Methylsulfonyl-1H-benzotriazole, 97%
1-Methylsulfonyl-1H-benzotriazole, 97%
1-Methylsulfonyl-1H-benzotriazole, 97%
Thermo Scientific Chemicals

1-Methylsulfonyl-1H-benzotriazole, 97%

CAS: 37073-15-7 | C7H7N3O2S | 197.21 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
25 g
Catalog number H53386.06
also known as H53386-06
Price (USD)
105.65
Special offer
Online exclusive
Ends: 30-Jun-2026
124.00
Save 18.35 (15%)
Each
Quantity:
5 g
Request bulk or custom format
Price (USD)
105.65
Special offer
Online exclusive
Ends: 30-Jun-2026
124.00
Save 18.35 (15%)
Each
Specifications
CAS37073-15-7
Chemical Name or Material1-Methylsulfonyl-1H-benzotriazole
Formula Weight197.21
Health Hazard 1H302-H318
Health Hazard 2GHS H Statement
H301-H318
Toxic if swallowed.
Causes serious eye damage.
View more
1-Methylsulfonyl-1H-benzotriazole is used as pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-Methylsulfonyl-1H-benzotriazole is used as pharmaceutical intermediate.

Notes
Stable under recommended storage conditions. Store away from oxidizing agents.
RUO – Research Use Only

General References:

  1. Alan R. Katritzky; Nabin K.Meher; Chunming Cai; Sandeep K.Singh. Preparation of N-acylbenzotriazole derivatives of dicarboxylic acids . Rev. Soc. Quim. Mex. 2004, 48, 275-278
  2. Alan R.Katritzky; Yuming Zhang; Sandeep K.Singh; Peter J.Steelb. 1,3-Dipolar cycloadditions of organic azides to ester or benzotriazolylcarbonyl activated acetylenic amides. ARKIVOC. 2003, (xv),47-64