1-Undecene, 97%
1-Undecene, 97%
1-Undecene, 97%
Thermo Scientific Chemicals

1-Undecene, 97%

CAS: 821-95-4 | C11H22 | 154.297 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Catalog number H53387.14
also known as H53387-14
Price (USD)
252.65
Online Exclusive
281.00
Save 28.35 (10%)
Each
Quantity:
25 g
Request bulk or custom format
Price (USD)
252.65
Online Exclusive
281.00
Save 28.35 (10%)
Each
Specifications
Beilstein1740044
CAS821-95-4
ChEBICHEBI:77444
Chemical Name or Material1-Undecene
Density0.751
View more
1-Undecene was used as a biomarker for identification of Pseudomonas aeruginosa strain. It is also used as pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-Undecene was used as a biomarker for identification of Pseudomonas aeruginosa strain. It is also used as pharmaceutical intermediate.

Solubility
Solunle in ether, chloroform and ligroin. Insoluble in water.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
RUO – Research Use Only

General References:

  1. Prof. Dr. Guosheng Liu; Guoyin Yin; Liang Wu. Palladium-catalyzed intermolecular aerobic oxidative amination of terminal alkenes: Efficient synthesis of linear allylamine d erivatives. Angewandte Chemie. 2008, 120, (25),4811-4814
  2. James M.Zechman; Scott Aldinger; John N.Labows Jr. Characterization of pathogenic bacteria by automated headspace concentration—gas chromatography. Journal of Chromatography B: Biomedical Sciences and Applications. 1986, 377, 49-57