tert-Butyl pivalate, 98%
tert-Butyl pivalate, 98%
tert-Butyl pivalate, 98%
Thermo Scientific Chemicals

tert-Butyl pivalate, 98%

CAS: 16474-43-4 | C9H18O2 | 158.24 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
100 g
Catalog number H53395.06
also known as H53395-06
Price (USD)
30.50
Each
Quantity:
5 g
Request bulk or custom format
Price (USD)
30.50
Each
Specifications
CAS16474-43-4
Chemical Name or Materialtert-Butyl pivalate
Density0.813
Flash Point>62°C (144°F)
Formula Weight158.24
View more
tert-Butyl pivalate is used as pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
tert-Butyl pivalate is used as pharmaceutical intermediate.

Notes
Stable under recommended storage conditions. Store away from oxidizing agents.
RUO – Research Use Only

General References:

  1. Satoru Masamune; Yoshio Hayase; Walter Schilling; Wan Kit Chan; Gordon S.Bates. Activation of thiol esters. Partial synthesis of cytochalasins A and B. J. Am. Chem. Soc. 1977, 99, (20),6756-6758
  2. Hanns Fischer; Henning Paul. Rate constants for some prototype radical reactions in liquids by kinetic electron spin resonance. Acc. Chem. Res. 1987, 20, (5),200-206