3-(Trimethylsilyl)propiolaldehyde diethyl acetal, 97%, Thermo Scientific Chemicals
3-(Trimethylsilyl)propiolaldehyde diethyl acetal, 97%, Thermo Scientific Chemicals
3-(Trimethylsilyl)propiolaldehyde diethyl acetal, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

3-(Trimethylsilyl)propiolaldehyde diethyl acetal, 97%, Thermo Scientific Chemicals

Have Questions?
Catalog NumberQuantity
H53397.06
also known as H53397-06
5 g
Catalog number H53397.06
also known as H53397-06
Price (USD)
60.40
Each
Quantity:
5 g
Request bulk or custom format
Price (USD)
60.40
Each
Specifications
Boiling Point170°C to 172°C
CAS87219-80-5
Chemical Name or Material3-(Trimethylsilyl)propiolaldehyde diethyl acetal
DOT InformationTransport Hazard Class: 3; Packing Group: III; Proper Shipping Name: FLAMMABLE LIQUIDS, N.O.S.
Flash Point40°C (104°F)
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only