2-Chloropyridine-3-carboxaldehyde, 98%
2-Chloropyridine-3-carboxaldehyde, 98%
2-Chloropyridine-3-carboxaldehyde, 98%
Thermo Scientific Chemicals

2-Chloropyridine-3-carboxaldehyde, 98%

CAS: 36404-88-3 | C6H4ClNO | 141.554 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
25 g
Catalog number H53399.06
also known as H53399-06
Price (USD)
106.65
Online Exclusive
118.00
Save 11.35 (10%)
Each
Quantity:
5 g
Request bulk or custom format
Price (USD)
106.65
Online Exclusive
118.00
Save 11.35 (10%)
Each
Chemical Identifiers
CAS36404-88-3
IUPAC Name2-chloropyridine-3-carbaldehyde
Molecular FormulaC6H4ClNO
InChI KeyKHPAGGHFIDLUMB-UHFFFAOYSA-N
SMILESClC1=NC=CC=C1C=O
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (GC)>97.5%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only