cis-5-Norbornene-endo-2,3-dicarboxylic acid, 98%, Thermo Scientific Chemicals
cis-5-Norbornene-endo-2,3-dicarboxylic acid, 98%, Thermo Scientific Chemicals
cis-5-Norbornene-endo-2,3-dicarboxylic acid, 98%, Thermo Scientific Chemicals
cis-5-Norbornene-endo-2,3-dicarboxylic acid, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

cis-5-Norbornene-endo-2,3-dicarboxylic acid, 98%, Thermo Scientific Chemicals

Have Questions?
Catalog NumberQuantity
H53403.14
also known as H53403-14
25 g
Catalog number H53403.14
also known as H53403-14
Price (USD)
139.00
Each
Add to cart
Quantity:
25 g
Request bulk or custom format
Price (USD)
139.00
Each
Add to cart
Specifications
CAS3853-88-1
Chemical Name or Materialcis-5-Norbornene-endo-2,3-dicarboxylic acid
Formula Weight182.18
Health Hazard 1H315-H319-H335
Health Hazard 2GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only