Cyclohexylacetylene, 98%
Cyclohexylacetylene, 98%
Cyclohexylacetylene, 98%
Thermo Scientific Chemicals

Cyclohexylacetylene, 98%

CAS: 931-48-6 | C8H12 | 108.184 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Catalog number H53418.06
also known as H53418-06
Price (USD)
119.65
Special offer
Online exclusive
Ends: 30-Jun-2026
140.00
Save 20.35 (15%)
Each
Quantity:
5 g
Request bulk or custom format
Price (USD)
119.65
Special offer
Online exclusive
Ends: 30-Jun-2026
140.00
Save 20.35 (15%)
Each
Chemical Identifiers
CAS931-48-6
IUPAC Nameethynylcyclohexane
Molecular FormulaC8H12
InChI KeySSDZYLQUYMOSAK-UHFFFAOYSA-N
SMILESC#CC1CCCCC1
View more
SpecificationsSpecification SheetSpecification Sheet
FormLiquid
Assay (GC)>97.5%
Appearance (Color)Colorless
Cyclohexylacetylene is used in the preparation of hydrido-vinylidene complexes of osmium, which finds vital intermediates in several homogeneous and heterogeneous catalytic reactions such as alkene olimerization, polymerization and Fischer-Tropsch synthesis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Cyclohexylacetylene is used in the preparation of hydrido-vinylidene complexes of osmium, which finds vital intermediates in several homogeneous and heterogeneous catalytic reactions such as alkene olimerization, polymerization and Fischer-Tropsch synthesis.

Solubility
Immiscible with water.

Notes
Incompatible with strong oxidizing agents.
RUO – Research Use Only

General References:

  1. Jacquet, J.; Auvinet, A. L.; Mandadapu, A. K.; Haddad, M.; Ratovelomanana-Vidal, V.; Michelet, V. Practical Solvent-Free Ruthenium Trichloride-Mediated Benzannulation Approach to Fused Functionalized Arenes. Adv. Synth. Catal. 2015, 357 (7), 1387-1392.
  2. Zhang, A. L.; Yang, L. W.; Yang, N. F.; Zhang, J. Synthesis of enantiopure C3-symmetric bulky trialkanolamines and their enantioselective inductivity during the alkynylation of aldehydes. J. Organomet. Chem. 2015, 775, 88-93.