Methyl 4-ethynylbenzoate, 97%
Methyl 4-ethynylbenzoate, 97%
Methyl 4-ethynylbenzoate, 97%
Thermo Scientific Chemicals

Methyl 4-ethynylbenzoate, 97%

CAS: 3034-86-4 | C10H8O2 | 160.17 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number H53419.03
also known as H53419-03
Price (USD)
130.65
Online Exclusive
145.00
Save 14.35 (10%)
Each
Quantity:
1 g
Request bulk or custom format
Price (USD)
130.65
Online Exclusive
145.00
Save 14.35 (10%)
Each
Chemical Identifiers
CAS3034-86-4
IUPAC Namemethyl 4-ethynylbenzoate
Molecular FormulaC10H8O2
InChI KeyJPGRSTBIEYGVNO-UHFFFAOYSA-N
SMILESCOC(=O)C1=CC=C(C=C1)C#C
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (HPLC)>96.0%
Methyl 4-ethynylbenzoate is used as pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Methyl 4-ethynylbenzoate is used as pharmaceutical intermediate.

Solubility
Sparingly soluble in water (0.16 g/L at 25°C).

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
RUO – Research Use Only

General References:

  1. Jognandan K.Prashar; Douglas E.Moore. Synthesis of carboranyl phenylalanine for potential use in neutron capture therapy of melanoma. J. Chem. Soc., Perkin Trans. 1. 1993, (9),1051-1053
  2. Ronald A.Forsch; Sherry F.Queener; Andre Rosowsky. Preliminary in vitro studies on two potent, water-soluble trimethoprim analogues with exceptional species selectivity against dihydrofolate reductase from Pneumocystis carinii and Mycobacterium avium. Bioorganic & Medicinal Chemistry Letters. 2004, 14, (7),1811-1815