3-Iodo-1-phenylpropane, 97%
3-Iodo-1-phenylpropane, 97%
3-Iodo-1-phenylpropane, 97%
Thermo Scientific Chemicals

3-Iodo-1-phenylpropane, 97%

CAS: 4119-41-9 | C9H11I | 246.09 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
25 g
Catalog number H53420.06
also known as H53420-06
Price (USD)
119.65
Special offer
Online exclusive
Ends: 30-Jun-2026
140.00
Save 20.35 (15%)
Each
Quantity:
5 g
Request bulk or custom format
Price (USD)
119.65
Special offer
Online exclusive
Ends: 30-Jun-2026
140.00
Save 20.35 (15%)
Each
Specifications
CAS4119-41-9
Chemical Name or Material3-Iodo-1-phenylpropane
Density1.53
Flash Point110°C (230°F)
Formula Weight246.09
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only