1-Phenyl-1-[4-(1-piperidinyl)phenyl]-2-propyn-1-ol, 97%
1-Phenyl-1-[4-(1-piperidinyl)phenyl]-2-propyn-1-ol, 97%
1-Phenyl-1-[4-(1-piperidinyl)phenyl]-2-propyn-1-ol, 97%
Thermo Scientific Chemicals

1-Phenyl-1-[4-(1-piperidinyl)phenyl]-2-propyn-1-ol, 97%

CAS: 214746-69-7 | C20H21NO | 291.394 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Catalog number H53435.06
also known as H53435-06
Price (USD)
70.65
Special offer
Online exclusive
Ends: 30-Jun-2026
82.80
Save 12.15 (15%)
Each
Quantity:
5 g
Request bulk or custom format
Price (USD)
70.65
Special offer
Online exclusive
Ends: 30-Jun-2026
82.80
Save 12.15 (15%)
Each
Specifications
CAS214746-69-7
Chemical Name or Material1-Phenyl-1-[4-(1-piperidinyl)phenyl]-2-propyn-1-ol
Formula Weight291.39
Health Hazard 1H315-H319-H335
Health Hazard 2GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
View more
1-Phenyl-1-[4-(1-piperidinyl)phenyl]-2-propyn-1-ol is used as an organic chemical synthesis intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-Phenyl-1-[4-(1-piperidinyl)phenyl]-2-propyn-1-ol is used as an organic chemical synthesis intermediate.

Notes
Stable under recommended storage conditions. Store away from oxidizing agents.
RUO – Research Use Only