4-Methyl-2-pentyne, 97%
4-Methyl-2-pentyne, 97%
4-Methyl-2-pentyne, 97%
Thermo Scientific Chemicals

4-Methyl-2-pentyne, 97%

CAS: 21020-27-9 | C6H10 | 82.15 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
25 g
Catalog number H53442.06
also known as H53442-06
Price (USD)
90.65
Special offer
Online exclusive
Ends: 30-Jun-2026
107.00
Save 16.35 (15%)
Each
Quantity:
5 g
Request bulk or custom format
Price (USD)
90.65
Special offer
Online exclusive
Ends: 30-Jun-2026
107.00
Save 16.35 (15%)
Each
Specifications
CAS21020-27-9
Chemical Name or Material4-Methyl-2-pentyne
DOT InformationTransport Hazard Class: 3; Packing Group: II; Proper Shipping Name: HYDROCARBONS, LIQUID, N.O.S.
Density0.711
Flash Point−12°C (−10°F)
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only