Allyloxytrimethylsilane, 98%
Allyloxytrimethylsilane, 98%
Allyloxytrimethylsilane, 98%
Thermo Scientific Chemicals

Allyloxytrimethylsilane, 98%

CAS: 18146-00-4 | C6H14OSi | 130.262 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Catalog number H53443.06
also known as H53443-06
Price (USD)
20.65
Special offer
Online exclusive
Ends: 30-Jun-2026
24.70
Save 4.05 (16%)
Each
Quantity:
5 g
Request bulk or custom format
Price (USD)
20.65
Special offer
Online exclusive
Ends: 30-Jun-2026
24.70
Save 4.05 (16%)
Each
Chemical Identifiers
CAS18146-00-4
IUPAC Nametrimethyl(prop-2-en-1-yloxy)silane
Molecular FormulaC6H14OSi
InChI KeyMNMVKGDEKPPREK-UHFFFAOYSA-N
SMILESC[Si](C)(C)OCC=C
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (GC)98.5% min
Allyloxytrimethylsilane is used in the synthesis of metalated dienes via an isomerization-elimination sequence. Allyloxytrimethylsilane [3-(Trimethylsiloxy)propene] was used in the synthesis of trimethylsilyl protected (3-hydroxypropyl)-trimethoxysilane. Used in preparation of electrical materials, OLED, and in Pharma industry.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Allyloxytrimethylsilane is used in the synthesis of metalated dienes via an isomerization-elimination sequence. Allyloxytrimethylsilane [3-(Trimethylsiloxy)propene] was used in the synthesis of trimethylsilyl protected (3-hydroxypropyl)-trimethoxysilane. Used in preparation of electrical materials, OLED, and in Pharma industry.

Notes
Moisture Sensitive.Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents, moisture.
RUO – Research Use Only

General References:

  1. John B Grande et. al. Rapid assembly of explicit, functional silicones.. Dalton Transactions. 2010, 39 (39), 9369-9378.
  2. Yasuyuki Tezuka,; Atsushi Okabayashi,; Kiyokazu Imai. Synthesis of poly(vinyl alcohol)-graft-poly(tetrahydrofuran). Die Makromolekulare Chemie. 1989 , 190 (4), 753-762.