cis-5-Norbornene-exo-2,3-dicarboxylic anhydride, 95%
cis-5-Norbornene-exo-2,3-dicarboxylic anhydride, 95%
cis-5-Norbornene-exo-2,3-dicarboxylic anhydride, 95%
Thermo Scientific Chemicals

cis-5-Norbornene-exo-2,3-dicarboxylic anhydride, 95%

CAS: 2746-19-2 | C9H8O3 | 164.16 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Catalog number H53446.06
also known as H53446-06
Price (USD)
176.65
Online Exclusive
196.00
Save 19.35 (10%)
Each
Quantity:
5 g
Request bulk or custom format
Price (USD)
176.65
Online Exclusive
196.00
Save 19.35 (10%)
Each
Chemical Identifiers
CAS2746-19-2
IUPAC Name(2R,6S)-4-oxatricyclo[5.2.1.0²,⁶]dec-8-ene-3,5-dione
Molecular FormulaC9H8O3
InChI KeyKNDQHSIWLOJIGP-CYDAGYADSA-N
SMILESO=C1OC(=O)[C@@H]2C3CC(C=C3)[C@H]12
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (GC)94.0% min
cis-5-Norbornene-exo-2,3-dicarboxylic anhydride is used as an organic chemical synthesis intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
cis-5-Norbornene-exo-2,3-dicarboxylic anhydride is used as an organic chemical synthesis intermediate.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
RUO – Research Use Only

General References:

  1. Boo-Gyo Shin; Myoung-Souk Jang; Do Yeung Yoon; Walter Heitz. Vinyl-type polymerization of norbornene dicarboxylic acid dialkyl esters . Macromolecular Rapid Communications. 2004, 25, (6),728-732
  2. Lixia Ren; Jiuyang Zhang; Xiaolong Bai; Christopher G.Hardy; Ken D.Shimizu; Chuanbing Tang. Preparation of cationic cobaltocenium polymers and block copolymers by living ring-opening metathesis polymerization. Chem. Sci. 2012, 3, (2),580-583