Tripropargylamine, 97%
Tripropargylamine, 97%
Tripropargylamine, 97%
Thermo Scientific Chemicals

Tripropargylamine, 97%

CAS: 6921-29-5 | C9H9N | 131.178 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
25 g
Catalog number H53459.03
also known as H53459-03
Price (USD)
44.65
Special offer
Online exclusive
Ends: 28-Dec-2026
51.80
Save 7.15
Each
Quantity:
1 g
Request bulk or custom format
Price (USD)
44.65
Special offer
Online exclusive
Ends: 28-Dec-2026
51.80
Save 7.15
Each
Specifications
CAS6921-29-5
Chemical Name or MaterialTripropargylamine
DOT InformationTransport Hazard Class: 3; Packing Group: III; Proper Shipping Name: FLAMMABLE LIQUIDS, N.O.S.
Density0.927
EINECS Number230-038-5
View more
Tripropargylamine is used as pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Tripropargylamine is used as pharmaceutical intermediate.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
RUO – Research Use Only

General References:

  1. Sachin A.Ingale; Frank Seela. A ratiometric fluorescent on-off Zn2+ chemosensor based on a tripropargylamine pyrene azide click adduct. J. Org. Chem. 2012, 77, (20),9352-9356
  2. Yeong-Soon Gal; Won-Chul Lee; Sam-Kwon Choi; Yun-Chul Kim; Bal Jung. Polymerization of tripropargylamine by transition metal catalysts and molecular structure of poly(tripropargylamine). European Polymer Journal. 1996, 32, (5),579-585