2-Hexene, cis + trans, tech. 85%
2-Hexene, cis + trans, tech. 85%
2-Hexene, cis + trans, tech. 85%
Thermo Scientific Chemicals

2-Hexene, cis + trans, tech. 85%

CAS: 592-43-8 | C6H12 | 84.162 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Catalog number H53472.06
also known as H53472-06
Price (USD)
70.65
Online exclusive
78.50
Save 7.85
Each
Quantity:
5 g
Request bulk or custom format
Price (USD)
70.65
Online exclusive
78.50
Save 7.85
Each
Specifications
Beilstein969179
CAS592-43-8
Chemical Name or Material2-Hexene
DOT InformationTransport Hazard Class: 3; Packing Group: II; Proper Shipping Name: HYDROCARBONS, LIQUID, N.O.S.
Density0.678
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only