2-(Trifluoromethyl)phenylacetylene, 97%
2-(Trifluoromethyl)phenylacetylene, 97%
2-(Trifluoromethyl)phenylacetylene, 97%
2-(Trifluoromethyl)phenylacetylene, 97%
Thermo Scientific Chemicals

2-(Trifluoromethyl)phenylacetylene, 97%

CAS: 704-41-6 | C9H5F3 | 170.134 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number H53478.06
also known as H53478-06
Price (USD)
185.65
Online Exclusive
206.00
Save 20.35 (10%)
Each
Add to cart
Quantity:
5 g
Request bulk or custom format
Price (USD)
185.65
Online Exclusive
206.00
Save 20.35 (10%)
Each
Add to cart
Specifications
CAS704-41-6
Chemical Name or Material2-(Trifluoromethyl)phenylacetylene
DOT InformationTransport Hazard Class: 3; Packing Group: III; Proper Shipping Name: FLAMMABLE LIQUIDS, N.O.S.
Density1.222
Flash Point25°C (77°F)
View more
2-(Trifluoromethyl)phenylacetylene is used as pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-(Trifluoromethyl)phenylacetylene is used as pharmaceutical intermediate.

Solubility
Sparingly soluble in water.

Notes
Stable under recommended storage conditions. Store away from oxidizing agents.
RUO – Research Use Only
Martin G. Mayershofer; Oskar Nuyken; Michael R.Buchmeiser. Bi- and trinuclear ruthenium alkylidene triggered cyclopolymerization of 1,6-heptadiynes:  Access to An-X-An block and (An)3X tristar copolymers. Macromolecules. 2006, 39, (10),3484-3493