tert-Butyl propargyl ether, 98%
tert-Butyl propargyl ether, 98%
tert-Butyl propargyl ether, 98%
tert-Butyl propargyl ether, 98%
Thermo Scientific Chemicals

tert-Butyl propargyl ether, 98%

CAS: 16314-18-4 | C7H12O | 112.17 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
100 g
Catalog number H53480.06
also known as H53480-06
Price (USD)
46.30
Each
Add to cart
Quantity:
5 g
Request bulk or custom format
Price (USD)
46.30
Each
Add to cart
Specifications
CAS16314-18-4
Chemical Name or Materialtert-Butyl propargyl ether
DOT InformationTransport Hazard Class: 3; Packing Group: II; Proper Shipping Name: FLAMMABLE LIQUIDS, N.O.S.
Density0.835
Flash Point20°C (68°F)
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only