Benzyl propargyl ether, 97%
Benzyl propargyl ether, 97%
Benzyl propargyl ether, 97%
Benzyl propargyl ether, 97%
Thermo Scientific Chemicals

Benzyl propargyl ether, 97%

CAS: 4039-82-1 | C10H10O | 146.189 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Catalog number H53481.14
also known as H53481-14
Price (USD)
145.65
Online Exclusive
162.00
Save 16.35 (10%)
Each
Add to cart
Quantity:
25 g
Request bulk or custom format
Price (USD)
145.65
Online Exclusive
162.00
Save 16.35 (10%)
Each
Add to cart
Specifications
CAS4039-82-1
Chemical Name or MaterialBenzyl propargyl ether
Density1.011
Formula Weight146.19
Health Hazard 1H315-H319-H335
View more
Benzyl propargyl ether is used in the synthesis of substituted carbocyclic aromatic compounds and pharmaceutical compounds.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Benzyl propargyl ether is used in the synthesis of substituted carbocyclic aromatic compounds and pharmaceutical compounds.

Solubility
Soluble in chloroform.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
RUO – Research Use Only

General References:

  1. Jae Wook Lee; Jung Hwan Kim; Byung-Ku Kim; Won Suk Shin; Sung-Ho Jin. Synthesis of Frechet type dendritic benzyl propargyl ether and Frechet type triazole dendrimer. Tetrahedron. 2006, 62, (5),894-900
  2. Benoit Bolte; Yann Odabachian; Fabien Gagosz. Gold(I)-catalyzed rearrangement of propargyl benzyl ethers: A practical method for the generation and in situ transformation of substituted allenes. J. Am. Chem. Soc. 2010, 132, (21),7294-7296