Benzophenone p-toluenesulfonylhydrazone, 97%
Benzophenone p-toluenesulfonylhydrazone, 97%
Benzophenone p-toluenesulfonylhydrazone, 97%
Thermo Scientific Chemicals

Benzophenone p-toluenesulfonylhydrazone, 97%

CAS: 4545-20-4 | C20H18N2O2S | 350.436 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Catalog number H53486.06
also known as H53486-06
Price (USD)
39.00
Each
Quantity:
5 g
Request bulk or custom format
Price (USD)
39.00
Each
Specifications
CAS4545-20-4
Chemical Name or MaterialBenzophenone p-toluenesulfonylhydrazone
Formula Weight350.44
IUPAC NameN-(benzhydrylideneamino)-4-methylbenzenesulfonamide
InChI KeyAVWHDUALELPLAI-UHFFFAOYSA-N
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only