N-(5-Hexynyl)phthalimide, 97%
N-(5-Hexynyl)phthalimide, 97%
N-(5-Hexynyl)phthalimide, 97%
Thermo Scientific Chemicals

N-(5-Hexynyl)phthalimide, 97%

CAS: 6097-08-1 | C14H13NO2 | 227.26 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Catalog number H53489.14
also known as H53489-14
Price (USD)
288.65
Online Exclusive
320.00
Save 31.35 (10%)
Each
Quantity:
25 g
Request bulk or custom format
Price (USD)
288.65
Online Exclusive
320.00
Save 31.35 (10%)
Each
Chemical Identifiers
CAS6097-08-1
IUPAC Name2-(hex-5-yn-1-yl)-2,3-dihydro-1H-isoindole-1,3-dione
Molecular FormulaC14H13NO2
InChI KeyWALVSJIWVWDFCU-UHFFFAOYSA-N
SMILESO=C1N(CCCCC#C)C(=O)C2=CC=CC=C12
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (unspecified)≥96.0%
6-amino-1-hexyne was prepared from N-(5-Hexynyl)phthalimide which is the starting material. Used in the synthesis of Amino-alkyne.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
6-amino-1-hexyne was prepared from N-(5-Hexynyl)phthalimide which is the starting material. Used in the synthesis of Amino-alkyne.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
RUO – Research Use Only

General References:

  1. Andreas Rennhack,; Thorsten Jumpertz,; Julia Ness,; Sandra Baches,; Claus U. Pietrzik,; Sascha Weggen,; Bruno Bulica. Synthesis of a potent photoreactive acidic γ-secretase modulator for target identification in cells. Bioorganic & Medicinal Chemistry. year of publication , 20 (21), 6523-6532.
  2. Sterne, Robert Synthesis of Novel Linear Multivalent Peptide Ligands Based on the Tetrapeptide MSH(4). J Master's Theses . 2010,