cis-2-Hexene is used as an organic chemical synthesis intermediate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
cis-2-Hexene is used as an organic chemical synthesis intermediate.
Solubility
Immiscible in water.
Notes
Stable under recommended storage conditions. Store away from oxidizing agents.
RUO – Research Use Only
General References:
- Richard R.Schrock; John A.Osborn. Catalytic hydrogenation using cationic rhodium complexes. II. The selective hydrogenation of alkynes to cis olefins. J. Am. Chem. Soc. 1976, 98, (8),2143-2147
- Giuliano Dolcetti. Olefins reduction by homogeneous nitrosyl catalysts. Inorganic and Nuclear Chemistry Letters. 1973, 9, (7),705-709