3-Chloro-1-phenyl-1-propyne, 97%
3-Chloro-1-phenyl-1-propyne, 97%
3-Chloro-1-phenyl-1-propyne, 97%
Thermo Scientific Chemicals

3-Chloro-1-phenyl-1-propyne, 97%

CAS: 3355-31-5 | C9H7Cl | 150.605 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
25 g
Catalog number H53510.06
also known as H53510-06
Price (USD)
104.65
Online Exclusive
116.00
Save 11.35 (10%)
Each
Quantity:
5 g
Request bulk or custom format
Price (USD)
104.65
Online Exclusive
116.00
Save 11.35 (10%)
Each
Specifications
CAS3355-31-5
Chemical Name or Material3-Chloro-1-phenyl-1-propyne
Density1.095
Flash Point104°C (219°F)
Formula Weight150.61
View more
3-Chloro-1-phenyl-1-propyne is used as an organic chemical synthesis intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3-Chloro-1-phenyl-1-propyne is used as an organic chemical synthesis intermediate.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
RUO – Research Use Only

General References:

  1. Yanchi Tang; Tiebo Xiao; Lei Zhou. Cobalt-catalyzed alkyne-dihalomethane-amine coupling: an efficient route for propargylamines. Tetrahedron Letters. 2012, 53, (46),6199-6201
  2. Herman G.RicheyJr.; L.Meredith Moses; M.S.Domalski; Wayne F.Erickson; Alan S.Heyn. Reactions of allylic Grignard reagents and unsaturated amines. J. Org. Chem. 1981, 46, (19),3773-3780