3,3'-Diaminobenzidine, 98+%
3,3'-Diaminobenzidine, 98+%
3,3'-Diaminobenzidine, 98+%
Thermo Scientific Chemicals

3,3'-Diaminobenzidine, 98+%

CAS: 91-95-2 | C12H14N4 | 214.272 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
100 g
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number H54000.06
also known as H54000-06
Price (USD)
-
Quantity:
5 g
Request bulk or custom format
Chemical Identifiers
CAS91-95-2
IUPAC Name[1,1'-biphenyl]-3,3',4,4'-tetramine
Molecular FormulaC12H14N4
InChI KeyHSTOKWSFWGCZMH-UHFFFAOYSA-N
SMILESNC1=CC=C(C=C1N)C1=CC=C(N)C(N)=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Off-white to brown
FormPowder
Assay (HPLC)≥98.0%
3,3'-Diaminobenzidine is used as peroxidase substrate and a reagent for spectrophotometric determination of selenium. DAB is used for the immunohistochemical staining of nucleic acids and proteins. As a dark brown dye, was used as an antibody-specific stain to identify the paired antibodies in breast tissue. It may be used for the synthesis of a useful, linear, Schiff-base coordination polymer.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3,3′-Diaminobenzidine is used as peroxidase substrate and a reagent for spectrophotometric determination of selenium. DAB is used for the immunohistochemical staining of nucleic acids and proteins. As a dark brown dye, was used as an antibody-specific stain to identify the paired antibodies in breast tissue. It may be used for the synthesis of a useful, linear, Schiff-base coordination polymer.

Solubility
Soluble in water (partly).

Notes
Light sensitive. Incompatible with oxidizing agents and heat.
RUO – Research Use Only

General References:

  1. N.H.Chua. Photooxidation of 3,3'-diaminobenzidine by blue-green algae and Chlamydomonas reinhardii. Biochimica et Biophysica Acta. 1972, 267, (1),179-189 
  2. Volker Herzog; H.Dariush Fahimi. A new sensitive colorimetric assay for peroxidase using 3,3'-diaminobenzidine as hydrogen donor. Analytical Biochemistry. 1973, 55, (2),554-562