5,7-Dichloro-1-indanone, 97%
5,7-Dichloro-1-indanone, 97%
5,7-Dichloro-1-indanone, 97%
5,7-Dichloro-1-indanone, 97%
Thermo Scientific Chemicals

5,7-Dichloro-1-indanone, 97%

CAS: 448193-94-0 | C9H6Cl2O | 201.05 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number H54007.06
also known as H54007-06
Price (USD)
316.65
Online Exclusive
352.00
Save 35.35 (10%)
Each
Add to cart
Quantity:
5 g
Request bulk or custom format
Price (USD)
316.65
Online Exclusive
352.00
Save 35.35 (10%)
Each
Add to cart
Specifications
CAS448193-94-0
Chemical Name or Material5,7-Dichloro-1-indanone
Formula Weight201.05
Health Hazard 1H302-H319
Health Hazard 2GHS H Statement
H301-H319
Toxic if swallowed.
Causes serious eye irritation.
View more
5,7-Dichloro-1-indanone is used as an organic chemical synthesis intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
5,7-Dichloro-1-indanone is used as an organic chemical synthesis intermediate.

Solubility
Slightly soluble in water.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
RUO – Research Use Only