4-Chloroquinoline-6-boronic acid pinacol ester, 96%
4-Chloroquinoline-6-boronic acid pinacol ester, 96%
4-Chloroquinoline-6-boronic acid pinacol ester, 96%
4-Chloroquinoline-6-boronic acid pinacol ester, 96%
Thermo Scientific Chemicals

4-Chloroquinoline-6-boronic acid pinacol ester, 96%

CAS: 1201844-73-6 | C15H17BClNO2 | 289.566 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 mg
1 g
Catalog number H54085.03
also known as H54085-03
Price (USD)
252.65
Special offer
Online exclusive
Ends: 30-Jun-2026
297.00
Save 44.35 (15%)
Each
Add to cart
Quantity:
1 g
Request bulk or custom format
Price (USD)
252.65
Special offer
Online exclusive
Ends: 30-Jun-2026
297.00
Save 44.35 (15%)
Each
Add to cart
Specifications
CAS1201844-73-6
Chemical Name or Material4-Chloroquinoline-6-boronic acid pinacol ester
Formula Weight289.56
IUPAC Name4-chloro-6-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)quinoline
InChI KeyANPKKHFSNJKOAM-UHFFFAOYSA-N
View more
4-Chloroquinoline-6-boronic acid pinacol ester is used as pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Chloroquinoline-6-boronic acid pinacol ester is used as pharmaceutical intermediate.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
RUO – Research Use Only
Masami Kuriyama; Seira Matsuo; Mina Shinozawa; Osamu Onomura. Ether-imidazolium carbenes for Suzuki-Miyaura cross-coupling of heteroaryl chlorides with aryl/heteroarylboron reagents. Org. Lett. 2013, 15, (11),2716-2719